Assay Details
| Gene Symbol: clockb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178295.2 | NP_840080.2 | 18 - 19 | 2094 | 89 | |
| XM_005168339.4 | XP_005168396.1 | 19 - 20 | 2478 | 89 | ||
| GenBank mRNA | AB087256.1 | - | 18 - 19 | 1998 | 89 | |
| BC162682.1 | - | 18 - 19 | 2084 | 89 | ||
More Information
Set Membership: |
|

clockb
Genomic Map