Assay Details
| Gene Symbol: clockb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178295.2 | NP_840080.2 | 19 - 20 | 2364 | 58 | |
| XM_005168339.4 | XP_005168396.1 | 20 - 21 | 2748 | 58 | ||
| GenBank mRNA | AB087256.1 | - | 19 - 20 | 2268 | 58 | |
| BC162682.1 | - | 19 - 20 | 2354 | 58 | ||
| CT669240.2 | - | 1 - 2 | 604 | 58 | ||
More Information
Set Membership: |
|

clockb
Genomic Map