Assay Details
| Gene Symbol: prl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181437.3 | NP_852102.2 | 3 - 4 | 314 | 70 | |
| GenBank mRNA | AY135149.1 | - | 3 - 4 | 310 | 70 | |
| BC092358.1 | - | 3 - 4 | 314 | 70 | ||
More Information
Set Membership: |
|

prl
Genomic Map