Assay Details
| Gene Symbol: anxa1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181759.1 | NP_861424.1 | 11 - 12 | 999 | 78 | |
| GenBank mRNA | AY178794.1 | - | 11 - 12 | 999 | 78 | |
| BC092685.1 | - | 12 - 13 | 1019 | 78 | ||
| BC164147.1 | - | 11 - 12 | 1002 | 78 | ||
More Information
Set Membership: |
|

anxa1b
Genomic Map