Assay Details
| Gene Symbol: anxa1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181759.1 | NP_861424.1 | 9 - 10 | 821 | 106 | |
| GenBank mRNA | AY178794.1 | - | 9 - 10 | 821 | 106 | |
| BC092685.1 | - | 10 - 11 | 841 | 106 | ||
| BC164147.1 | - | 9 - 10 | 824 | 106 | ||
More Information
Set Membership: |
|

anxa1b
Genomic Map