Assay Details
| Gene Symbol: mknk2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194402.1 | NP_919383.1 | 4 - 5 | 473 | 88 | |
| GenBank mRNA | BC044375.1 | - | 4 - 5 | 473 | 88 | |
| BC165746.1 | - | 3 - 4 | 277 | 88 | ||
More Information
Set Membership: |
|

mknk2b
Genomic Map