Assay Details
| Gene Symbol: timm17a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198367.1 | NP_938181.1 | 5 - 6 | 429 | 72 | |
| GenBank mRNA | BC056525.1 | - | 5 - 6 | 429 | 72 | |
| BC065901.1 | - | 5 - 6 | 440 | 72 | ||
More Information
Set Membership: |
|

timm17a
Genomic Map