Assay Details
| Gene Symbol: mibp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198877.1 | NP_942578.1 | 5 - 6 | 627 | 93 | |
| GenBank mRNA | BC047858.1 | - | 5 - 6 | 627 | 93 | |
| BC066586.1 | - | 5 - 6 | 575 | 93 | ||
More Information
Set Membership: |
|

mibp2
Genomic Map