Assay Details
| Gene Symbol: glud1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199545.4 | NP_955839.2 | 12 - 13 | 1725 | 86 | |
| GenBank mRNA | AY577003.1 | - | 12 - 13 | 1725 | 86 | |
| BC044202.1 | - | 12 - 13 | 1661 | 86 | ||
| BC063940.1 | - | 12 - 13 | 1589 | 86 | ||
| BC165763.1 | - | 12 - 13 | 1543 | 86 | ||
More Information
Set Membership: |
|

glud1b
Genomic Map