Assay Details
| Gene Symbol: glud1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199545.4 | NP_955839.2 | 5 - 6 | 904 | 112 | |
| GenBank mRNA | AY577003.1 | - | 5 - 6 | 904 | 112 | |
| BC063940.1 | - | 5 - 6 | 768 | 112 | ||
More Information
Set Membership: |
|

glud1b
Genomic Map