Assay Details
| Gene Symbol: hsd17b12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199613.1 | NP_955907.1 | 7 - 8 | 636 | 152 | |
| GenBank mRNA | AY551080.1 | - | 7 - 8 | 526 | 152 | |
| BC059617.1 | - | 7 - 8 | 636 | 152 | ||
| BC152213.1 | - | 7 - 8 | 645 | 152 | ||
| BC154318.1 | - | 7 - 8 | 608 | 152 | ||
More Information
Set Membership: |
|

hsd17b12b
Genomic Map