Assay Details
| Gene Symbol: hsd17b12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199613.1 | NP_955907.1 | 9 - 10 | 785 | 79 | |
| GenBank mRNA | AY551080.1 | - | 9 - 10 | 79 | ||
| BC059617.1 | - | 9 - 10 | 785 | 79 | ||
| BC152213.1 | - | 9 - 10 | 79 | |||
| BC154318.1 | - | 9 - 10 | 79 | |||
More Information
Set Membership: |
|

hsd17b12b
Genomic Map