Assay Details
| Gene Symbol: upb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199616.1 | NP_955910.1 | 9 - 10 | 1102 | 66 | |
| GenBank mRNA | BC053204.1 | - | 9 - 10 | 1102 | 66 | |
| BC164705.1 | - | 9 - 10 | 1112 | 66 | ||
More Information
Set Membership: |
|

upb1
Genomic Map