Assay Details
| Gene Symbol: waca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199660.1 | NP_955954.1 | 12 - 13 | 1734 | 119 | |
| XM_005156197.2 | XP_005156254.1 | 12 - 13 | 1827 | 119 | ||
| XM_009306588.2 | XP_009304863.1 | 12 - 13 | 1830 | 119 | ||
| GenBank mRNA | AY576994.1 | - | 12 - 13 | 1736 | 119 | |
| BC048888.1 | - | 12 - 13 | 1734 | 119 | ||
More Information
Set Membership: |
|

waca
Genomic Map