Assay Details
| Gene Symbol: stk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199672.1 | NP_955966.1 | 9 - 10 | 1267 | 70 | |
| GenBank mRNA | BC048033.1 | - | 9 - 10 | 1267 | 70 | |
| BC164215.1 | - | 9 - 10 | 1183 | 70 | ||
More Information
Set Membership: |
|

stk3
Genomic Map