Assay Details
| Gene Symbol: tpm4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199675.2 | NP_955969.1 | 1 - 2 | 361 | 69 | |
| GenBank mRNA | BC055225.1 | - | 1 - 2 | 257 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

tpm4b| Gene Symbol: tpm4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199675.2 | NP_955969.1 | 1 - 2 | 361 | 69 | |
| GenBank mRNA | BC055225.1 | - | 1 - 2 | 257 | 69 | |
Set Membership: |
|