Assay Details
| Gene Symbol: tpm4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199675.2 | NP_955969.1 | 5 - 6 | 761 | 106 | |
| XM_017357672.2 | XP_017213161.1 | 1008 | 106 | |||
| XM_017357674.2 | XP_017213163.1 | 1008 | 106 | |||
| GenBank mRNA | AI444323.1 | - | 1 - 2 | 75 | 106 | |
| BC055225.1 | - | 5 - 6 | 657 | 106 | ||
| KY127425.1 | - | 5 - 6 | 651 | 106 | ||
More Information
Set Membership: |
|

tpm4b
Genomic Map