Assay Details
| Gene Symbol: tpm4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199675.2 | NP_955969.1 | 7 - 8 | 891 | 69 | |
| XM_017357672.2 | XP_017213161.1 | 1138 | 69 | |||
| GenBank mRNA | AI444323.1 | - | 3 - 4 | 205 | 69 | |
| BC055225.1 | - | 7 - 8 | 787 | 69 | ||
More Information
Set Membership: |
|

tpm4b
Genomic Map