Assay Details
| Gene Symbol: ccnt2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193381.1 | NP_001180310.1 | 5 - 6 | 543 | 118 | |
| NM_199828.2 | NP_956122.1 | 5 - 6 | 543 | 118 | ||
| GenBank mRNA | BC044378.1 | - | 5 - 6 | 543 | 118 | |
| BM776488.1 | - | 5 - 6 | 500 | 118 | ||
More Information
Set Membership: |
|

ccnt2a
Genomic Map