Assay Details
| Gene Symbol: ccnt2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193381.1 | NP_001180310.1 | 7 - 8 | 760 | 89 | |
| NM_199828.2 | NP_956122.1 | 7 - 8 | 760 | 89 | ||
| XM_021478610.1 | XP_021334285.1 | 4 - 5 | 405 | 89 | ||
| XM_021478611.1 | XP_021334286.1 | 2 - 3 | 505 | 89 | ||
| GenBank mRNA | BC044378.1 | - | 7 - 8 | 760 | 89 | |
| EB771057.1 | - | 2 - 3 | 195 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021478611.1 transcripts.
Set Membership: |
|

ccnt2a
Genomic Map