Assay Details
| Gene Symbol: ccnt2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199828.2 | NP_956122.1 | 9 - 10 | 1874 | 91 | |
| GenBank mRNA | BC044378.1 | - | 9 - 10 | 1874 | 91 | |
| EH526493.1 | - | 1 - 2 | 295 | 91 | ||
More Information
Set Membership: |
|

ccnt2a
Genomic Map