Assay Details
| Gene Symbol: rps12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282177.1 | NP_001269106.1 | 4 - 5 | 307 | 63 | |
| NM_200046.3 | NP_956340.1 | 3 - 4 | 436 | 63 | ||
| GenBank mRNA | AY561510.1 | - | 3 - 4 | 403 | 63 | |
| BC059433.1 | - | 3 - 4 | 279 | 63 | ||
| BI326372.1 | - | 3 - 4 | 257 | 63 | ||
| CT698501.2 | - | 4 - 5 | 308 | 63 | ||
More Information
Set Membership: |
|

rps12
Genomic Map