Assay Details
| Gene Symbol: rps6kal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200073.1 | NP_956367.1 | 4 - 5 | 439 | 101 | |
| GenBank mRNA | BC045478.1 | - | 4 - 5 | 543 | 101 | |
| BC057467.1 | - | 4 - 5 | 439 | 101 | ||
More Information
Set Membership: |
|

rps6kal
Genomic Map