Assay Details
| Gene Symbol: mocs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200127.1 | NP_956421.1 | 2 - 3 | 373 | 89 | |
| GenBank mRNA | BC042324.1 | - | 2 - 3 | 373 | 89 | |
| BC071459.1 | - | 2 - 3 | 335 | 89 | ||
| BC164541.1 | - | 2 - 3 | 255 | 89 | ||
More Information
Set Membership: |
|

mocs3
Genomic Map