Assay Details
| Gene Symbol: mocs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200127.1 | NP_956421.1 | 7 - 8 | 892 | 66 | |
| XM_021467123.1 | XP_021322798.1 | 4 - 5 | 381 | 66 | ||
| GenBank mRNA | BC042324.1 | - | 7 - 8 | 892 | 66 | |
| BC164541.1 | - | 7 - 8 | 774 | 66 | ||
More Information
Set Membership: |
|

mocs3
Genomic Map