Assay Details
| Gene Symbol: dnajb6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200305.1 | NP_956599.1 | 5 - 6 | 633 | 106 | |
| XM_009303365.3 | XP_009301640.1 | 5 - 6 | 703 | 106 | ||
| XM_009303366.2 | XP_009301641.1 | 5 - 6 | 718 | 106 | ||
| XM_021477435.1 | XP_021333110.1 | 5 - 6 | 718 | 106 | ||
| GenBank mRNA | BC049536.1 | - | 5 - 6 | 633 | 106 | |
More Information
Set Membership: |
|

dnajb6b
Genomic Map