Assay Details
| Gene Symbol: tmem54a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200347.1 | NP_956641.1 | 2 - 3 | 275 | 120 | |
| XM_005158499.4 | XP_005158556.1 | 3 - 4 | 120 | |||
| GenBank mRNA | BC053159.1 | - | 2 - 3 | 275 | 120 | |
More Information
Set Membership: |
|

tmem54a
Genomic Map