Assay Details
| Gene Symbol: psmd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200572.1 | NP_956866.1 | 2 - 3 | 434 | 52 | |
| GenBank mRNA | BC056598.1 | - | 2 - 3 | 434 | 52 | |
| BC066451.1 | - | 2 - 3 | 429 | 52 | ||
More Information
Set Membership: |
|

psmd3
Genomic Map