Assay Details
| Gene Symbol: wars1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200772.1 | NP_957066.1 | 1152 | 60 | ||
| GenBank mRNA | BC059603.1 | - | 1152 | 60 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

wars1| Gene Symbol: wars1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200772.1 | NP_957066.1 | 1152 | 60 | ||
| GenBank mRNA | BC059603.1 | - | 1152 | 60 | ||
Set Membership: |
|