Assay Details
| Gene Symbol: rom1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201014.2 | NP_957308.2 | 5 - 6 | 921 | 61 | |
| GenBank mRNA | BC049482.1 | - | 5 - 6 | 919 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

rom1b| Gene Symbol: rom1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201014.2 | NP_957308.2 | 5 - 6 | 921 | 61 | |
| GenBank mRNA | BC049482.1 | - | 5 - 6 | 919 | 61 | |
Set Membership: |
|