Assay Details
| Gene Symbol: spag7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201125.1 | NP_957419.1 | 3 - 4 | 257 | 56 | |
| GenBank mRNA | BC054697.1 | - | 3 - 4 | 257 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

spag7| Gene Symbol: spag7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201125.1 | NP_957419.1 | 3 - 4 | 257 | 56 | |
| GenBank mRNA | BC054697.1 | - | 3 - 4 | 257 | 56 | |
Set Membership: |
|