Assay Details
| Gene Symbol: rapgef1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328346.1 | NP_001315275.1 | 3 - 4 | 574 | 100 | |
| XM_021468780.1 | XP_021324455.1 | 3 - 4 | 697 | 100 | ||
| XM_021468781.1 | XP_021324456.1 | 3 - 4 | 697 | 100 | ||
| XM_021468782.1 | XP_021324457.1 | 3 - 4 | 695 | 100 | ||
| XM_021468783.1 | XP_021324458.1 | 3 - 4 | 696 | 100 | ||
| XM_021468784.1 | XP_021324459.1 | 3 - 4 | 694 | 100 | ||
| GenBank mRNA | BC049305.1 | - | 3 - 4 | 564 | 100 | |
More Information
Set Membership: |
|

rapgef1b
Genomic Map