Assay Details
| Gene Symbol: rapgef1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021468782.1 | XP_021324457.1 | 6 - 7 | 1099 | 135 | |
| GenBank mRNA | BC049305.1 | - | 6 - 7 | 968 | 135 | |
More Information
Set Membership: |
|

rapgef1b
Genomic Map