Assay Details
| Gene Symbol: rapgef1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021468781.1 | XP_021324456.1 | 8 - 9 | 99 | ||
| GenBank mRNA | BC049305.1 | - | 7 - 8 | 1080 | 99 | |
More Information
Set Membership: |
|

rapgef1b
Genomic Map