Assay Details
| Gene Symbol: oxsr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201175.3 | NP_957469.2 | 14 - 15 | 1446 | 84 | |
| GenBank mRNA | BC044183.1 | - | 14 - 15 | 1432 | 84 | |
| BC067372.1 | - | 14 - 15 | 1424 | 84 | ||
| BC165393.1 | - | 14 - 15 | 1330 | 84 | ||
| EH602240.1 | - | 4 - 5 | 496 | 84 | ||
More Information
Set Membership: |
|

oxsr1b
Genomic Map