Assay Details
| Gene Symbol: oxsr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201175.3 | NP_957469.2 | 15 - 16 | 1509 | 127 | |
| GenBank mRNA | BC067372.1 | - | 15 - 16 | 1487 | 127 | |
| BC165393.1 | - | 15 - 16 | 1393 | 127 | ||
| EH602240.1 | - | 3 - 4 | 433 | 127 | ||
More Information
Set Membership: |
|

oxsr1b
Genomic Map