Assay Details
| Gene Symbol: oxsr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201175.3 | NP_957469.2 | 16 - 17 | 1568 | 115 | |
| GenBank mRNA | BC044183.1 | - | 16 - 17 | 1554 | 115 | |
| BC067372.1 | - | 16 - 17 | 1546 | 115 | ||
| BC165393.1 | - | 16 - 17 | 1452 | 115 | ||
| EH602240.1 | - | 2 - 3 | 374 | 115 | ||
More Information
Set Membership: |
|

oxsr1b
Genomic Map