Assay Details
| Gene Symbol: oxsr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201175.3 | NP_957469.2 | 17 - 18 | 1641 | 144 | |
| GenBank mRNA | BC044183.1 | - | 17 - 18 | 1627 | 144 | |
| BC067372.1 | - | 17 - 18 | 1619 | 144 | ||
| BC165393.1 | - | 17 - 18 | 1525 | 144 | ||
| EH602240.1 | - | 1 - 2 | 301 | 144 | ||
More Information
Set Membership: |
|

oxsr1b
Genomic Map