Assay Details
| Gene Symbol: oxsr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201175.3 | NP_957469.2 | 6 - 7 | 747 | 87 | |
| XM_021470117.1 | XP_021325792.1 | 6 - 7 | 774 | 87 | ||
| GenBank mRNA | BC044183.1 | - | 6 - 7 | 733 | 87 | |
| BC067372.1 | - | 6 - 7 | 725 | 87 | ||
| BC165393.1 | - | 6 - 7 | 631 | 87 | ||
| EH577479.1 | - | 5 - 6 | 495 | 87 | ||
More Information
Set Membership: |
|

oxsr1b
Genomic Map