Assay Details
| Gene Symbol: oxsr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201175.3 | NP_957469.2 | 7 - 8 | 855 | 71 | |
| XM_021470117.1 | XP_021325792.1 | 7 - 8 | 882 | 71 | ||
| GenBank mRNA | BC044183.1 | - | 7 - 8 | 841 | 71 | |
| BC067372.1 | - | 7 - 8 | 833 | 71 | ||
| BC165393.1 | - | 7 - 8 | 739 | 71 | ||
| EH577479.1 | - | 4 - 5 | 387 | 71 | ||
More Information
Set Membership: |
|

oxsr1b
Genomic Map