Assay Details
| Gene Symbol: oxsr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201175.3 | NP_957469.2 | 9 - 10 | 1032 | 99 | |
| XM_021470117.1 | XP_021325792.1 | 9 - 10 | 1059 | 99 | ||
| GenBank mRNA | BC044183.1 | - | 9 - 10 | 1018 | 99 | |
| BC067372.1 | - | 9 - 10 | 1010 | 99 | ||
| BC165393.1 | - | 9 - 10 | 916 | 99 | ||
| EH577479.1 | - | 2 - 3 | 210 | 99 | ||
More Information
Set Membership: |
|

oxsr1b
Genomic Map