Assay Details
| Gene Symbol: cd151l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205650.1 | NP_991213.1 | 3 - 4 | 395 | 79 | |
| XM_009293634.3 | XP_009291909.1 | 3 - 4 | 341 | 79 | ||
| GenBank mRNA | BC065859.1 | - | 3 - 4 | 395 | 79 | |
More Information
Set Membership: |
|

cd151l
Genomic Map