Assay Details
| Gene Symbol: abhd14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207066.2 | NP_996949.2 | 4 - 5 | 750 | 73 | |
| GenBank mRNA | BC066291.1 | - | 4 - 5 | 701 | 73 | |
| EH433068.1 | - | 4 - 5 | 767 | 73 | ||
| EH459072.1 | - | 1 - 2 | 558 | 73 | ||
More Information
Set Membership: |
|

abhd14a
Genomic Map