Assay Details
| Gene Symbol: cd9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212619.1 | NP_997784.1 | 1 - 2 | 74 | 87 | |
| GenBank mRNA | BC059691.1 | - | 1 - 2 | 187 | 87 | |
| BC067597.1 | - | 1 - 2 | 74 | 87 | ||
More Information
Set Membership: |
|

cd9a
Genomic Map