Assay Details
| Gene Symbol: cd9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212619.1 | NP_997784.1 | 7 - 8 | 631 | 69 | |
| XM_005158981.4 | XP_005159038.1 | 7 - 8 | 1153 | 69 | ||
| GenBank mRNA | BC059691.1 | - | 7 - 8 | 744 | 69 | |
| BC067597.1 | - | 7 - 8 | 631 | 69 | ||
More Information
Set Membership: |
|

cd9a
Genomic Map