Assay Details
| Gene Symbol: atp6v0a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212672.1 | NP_997837.1 | 6 - 7 | 581 | 80 | |
| XM_005163798.3 | XP_005163855.1 | 6 - 7 | 693 | 80 | ||
| GenBank mRNA | BC066692.1 | - | 6 - 7 | 581 | 80 | |
More Information
Set Membership: |
|

atp6v0a1a
Genomic Map