Assay Details
| Gene Symbol: dpf2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212696.2 | NP_997861.2 | 1 - 2 | 76 | 89 | |
| GenBank mRNA | BC092130.1 | - | 1 - 2 | 76 | 89 | |
| BC165789.1 | - | 1 - 2 | 70 | 89 | ||
More Information
Set Membership: |
|

dpf2l
Genomic Map