Assay Details
| Gene Symbol: dpf2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212696.2 | NP_997861.2 | 4 - 5 | 538 | 110 | |
| GenBank mRNA | AY391455.1 | - | 4 - 5 | 488 | 110 | |
| BC092130.1 | - | 4 - 5 | 538 | 110 | ||
| BC165789.1 | - | 4 - 5 | 532 | 110 | ||
More Information
Set Membership: |
|

dpf2l
Genomic Map