Assay Details
| Gene Symbol: mpzl2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212763.2 | NP_997928.2 | 3 - 4 | 607 | 68 | |
| GenBank mRNA | AJ491253.1 | - | 3 - 4 | 611 | 68 | |
| BC075940.1 | - | 3 - 4 | 592 | 68 | ||
| BC092883.1 | - | 3 - 4 | 589 | 68 | ||
| CU458934.1 | - | 3 - 4 | 608 | 68 | ||
| EH448751.1 | - | 3 - 4 | 864 | 68 | ||
| EH517304.1 | - | 1 - 2 | 477 | 68 | ||
More Information
Set Membership: |
|

mpzl2b
Genomic Map