Assay Details
| Gene Symbol: chst11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212824.2 | NP_997989.2 | 1 - 2 | 202 | 123 | |
| GenBank mRNA | AB097217.1 | - | 1 - 2 | 195 | 123 | |
| BC124140.1 | - | 1 - 2 | 202 | 123 | ||
| BC164169.1 | - | 1 - 2 | 146 | 123 | ||
More Information
Set Membership: |
|

chst11
Genomic Map